Learn More
HTR3B Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HTR3B.
Supplier: Abnova Corporation PAB18713
Description
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. [provided by RefSeq
Sequence: C-SNYLQTQDQTDQQESpecifications
HTR3B | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
HTR3B | |
HTR3B | |
A synthetic peptide corresponding to amino acids near C-terminus of human HTR3B. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of HTR3B. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
5-HT3B | |
Goat | |
Antigen affinity purification | |
RUO | |
9177 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only