Learn More
Goat anti-Human Complement C3 Polyclonal Anitbody, HRP, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Goat
Species Reactivity: Human
Specifications
Specifications
| Antigen | Complement C3 |
| Classification | Polyclonal |
| Conjugate | HRP |
| Host Species | Goat |
| Quantity | 2 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Purified |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.