Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgG (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP260658 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260658 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260658 Supplier Novus Biologicals Supplier No. NBP260658
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot.

Specifications

Antigen IgG (H+L)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Dot Blot
Classification Polyclonal
Concentration 2.22mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Flow Cytometry, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000-1:5000, Dot Blot
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2; Stabilizer: 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free. with 0.05% Sodium Azide
Host Species Goat
Immunogen Goat anti-Human IgG (H+L) Secondary Antibody was produced by repeated immunization with Human IgG whole molecule in goat.
Purification Method Affinity Purified
Quantity 2 mg
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Goat Serum, Human IgG, and Human Serum.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.