Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human Lambda Light Chain Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP173604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173604 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173604 Supplier Novus Biologicals Supplier No. NBP173604

Goat Polyclonal Antibody

Lambda Light Chain Polyclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.

Specifications

Antigen Lambda Light Chain
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Fluorescein
Dilution Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Gene Alias IGLC, immunoglobulin lambda constant group
Gene Symbols IGLC1
Host Species Goat
Immunogen Human lambda free chain
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3536
Test Specificity Specificity was confirmed by ELISA minimal cross reactivity against other human heavy or light chain isotypes.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.