Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL1 Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133133 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-133 Supplier Abnova Corporation Supplier No. PAB7309

Goat polyclonal antibody raised against synthetic peptide of Hyal1.

Sequence: SDKTSTKESCQ

Specifications

Antigen HYAL1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description hyaluronoglucosaminidase 1
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Hyal1
Gene Alias Hya1, Hyal-1
Gene Symbols Hyal1
Host Species Goat
Immunogen A synthetic peptide corresponding to mouse Hyal1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15586
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.