Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxynonenal, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137566 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-566 Supplier Abnova Corporation Supplier No. PAB1294

Goat polyclonal antibody raised against hydroxynonenal.

Specifications

Antigen Hydroxynonenal
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against hydroxynonenal.
Dilution Western Blot (1:2000-1:8000) ELISA (1:2000-1:8000) The optimal working dilution should be determined by the end user.
Formulation In 75mM PBS, 75mM NaCl, 0.5mM EDTA, pH 7.2 (0.02% sodium azide)
Host Species Goat
Immunogen Hydroxynonenal conjugated with KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody specifically binds to HNE modified proteins.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.