Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia trachomatis, Elementary Bodies, Goat, FITC, Polyclonal Antibody, Abnova™

Catalog No. 89137850 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-850 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-850 Supplier Abnova Corporation Supplier No. PAB14211
Add to Cart
Add to Cart

Goat polyclonal antibody raised against Chlamydia trachomatis, Elementary Bodies.

Specifications

Antigen Chlamydia trachomatis, Elementary Bodies
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Description Goat polyclonal antibody raised against Chlamydia trachomatis, Elementary Bodies.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Host Species Goat
Immunogen L2 and other serovar groups.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Purified elementary bodies, disrupted. Cross-reacts with Chlamydia psittacii & Chlamydia pneumoniae (TWAR). Negative against HEp-2 cells and egg yolk sac. All antigens of strain L2.
Target Species Bacteria
Content And Storage Store in the dark at 4°C. For long term storage store at -20°C.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.