Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia trachomatis, Major Outer Membrane Protein, Goat, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89137855 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-855 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-855 Supplier Abnova Corporation Supplier No. PAB14216

Goat polyclonal antibody raised against Chlamydia trachomatis, Major Outer Membrane Protein.

Specifications

Antigen Chlamydia trachomatis, Major Outer Membrane Protein
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation In PBS (10mg/mL BSA, 0.002% thimerosal)
Host Species Goat
Immunogen Native purified MOMP from strain L2.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Major Outer Membrane Protein (MOMP). Uninfected Cell Reactivity is negative against HEp-2 cells and egg yolk sac. Does not react with C. psittacii or C. pneumoniae in MIF.
Target Species Bacteria
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.