Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum D Toxoid, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89136918 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-918 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-918 Supplier Abnova Corporation Supplier No. PAB0915

Goat polyclonal antibody raised against Clostridium botulinum D Toxoid.

Specifications

Antigen Clostridium botulinum D Toxoid
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against Clostridium botulinum D Toxoid.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Goat
Immunogen Clostridium botulinum D Toxoid.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with Clostridium botulinum D Toxoid
Target Species Clostridium species
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.