Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Escherichia coli (O/K antigens), Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against Escherichia coli (O/K antigens).
Supplier: Abnova Corporation PAB13940
Specifications
Escherichia coli (O/K antigens) | |
Polyclonal | |
Goat polyclonal antibody raised against Escherichia coli (O/K antigens). | |
In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
Mixture of Escherichia coli serotypes. | |
RUO | |
Many “O” and “K” antigenic serotypes of Escherichia coli. Antiserum is not absorbed and does react with related Enterobacteriaceae. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Goat | |
1 mL | |
Primary | |
E. coli | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction