Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter pylori antigens, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89138617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-617 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-617 Supplier Abnova Corporation Supplier No. PAB8030
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native Helicobacter pylori.

Specifications

Antigen Helicobacter pylori antigens
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native Helicobacter pylori.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide)
Host Species Goat
Immunogen Native purified Helicobacter pylori antigens.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species H. pylori
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.