Learn More
Goat anti-immunodeficiency virus type 1 (HIV-1), Biotin, Polyclonal, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | immunodeficiency virus type 1 (HIV-1) |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | In 10mM PBS, pH 7.2 (0.09% sodium azide) |
| Host Species | Goat |
| Immunogen | Native human immunodeficiency virus type 1. |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Target Species | Virus |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.