Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

immunodeficiency virus type 1 (HIV-1) gp120, Goat, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89137899 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-899 Supplier Abnova Corporation Supplier No. PAB14269

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Goat polyclonal antibody raised against human immunodeficiency virus type 1 gp120.

Specifications

Antigen immunodeficiency virus type 1 (HIV-1) gp120
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Description Goat polyclonal antibody raised against human immunodeficiency virus type 1 gp120.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (10mg/mL BSA, 0.002% thimerosal)
Host Species Goat
Immunogen Native purified native gp120 from strain IIIB.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity gp120 (predominate gp120 band in Western blot). Does not cross-react with human T or B cells. Not reactive with human or bovine sera.
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.