Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

immunodeficiency virus type 1 (HIV-1) p24, Goat, FITC, Polyclonal Antibody, Abnova™

Catalog No. 89137901 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-901 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-901 Supplier Abnova Corporation Supplier No. PAB14271
Add to Cart
Add to Cart

Goat polyclonal antibody raised against human immunodeficiency virus type 1 p24.

Specifications

Antigen immunodeficiency virus type 1 (HIV-1) p24
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate FITC
Description Goat polyclonal antibody raised against human immunodeficiency virus type 1 p24.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Host Species Goat
Immunogen Native purified native p24 from strain IIIB.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity p24 (predominate p24 band pattern in Western blot). Antiserum does not cross-react with human T or B cells. Antiserum is not reactive with human or bovine sera.
Target Species Virus
Content And Storage Store in the dark at 4°C. For long term storage store at -20°C.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.