Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Strain A/USSR/90/77 (H1N1), Goat, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89137908 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-908 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-908 Supplier Abnova Corporation Supplier No. PAB14278
Add to Cart
Add to Cart

Goat polyclonal antibody raised against Influenza A Strain A/USSR/90/77 (H1N1)..

Specifications

Antigen Influenza A Strain A/USSR/90/77 (H1N1)
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate HRP
Description Goat polyclonal antibody raised against Influenza A Strain A/USSR/90/77 (H1N1)..
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (10mg/mL BSA, 0.002% thimerosal)
Host Species Goat
Immunogen Influenza A Strain A/USSR/90/77 (H1N1).
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Purified virions. Specific to H1N1 by IHA. Reacts with all antigenic types of Influenza A in other assay applications. Does not react Inf. B, RSV, Para 1-3 or Adeno. Does not react with HEp-2 cells. May react with chicken cellular proteins.
Target Species Influenza virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.