Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A, strain Texas 1/77 (H3N2), Goat, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89137911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-911 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-911 Supplier Abnova Corporation Supplier No. PAB14281
Add to Cart
Add to Cart

Goat polyclonal antibody raised against Influenza A, strain Texas 1/77 (H3N2).

Specifications

Antigen Influenza A, strain Texas 1/77 (H3N2)
Applications ELISA
Classification Polyclonal
Conjugate HRP
Description Goat polyclonal antibody raised against Influenza A, strain Texas 1/77 (H3N2).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (10mg/mL BSA, 0.002% thimerosal)
Host Species Goat
Immunogen Influenza A Strain Texas 1/77 (H3N2).
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Purified virions. Reacts with all antigenic types of Influenza A in other assay applications. May react with chicken cellular proteins. Specific to H3N2 by IHA. Does not react with HEp-2 cells. Does not react Influenza B, RSV, Para 1-3 or Adenovirus.
Target Species Influenza virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.