Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B (virions), Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137785 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-785 Supplier Abnova Corporation Supplier No. PAB13993

Goat polyclonal antibody raised against Influenza B (virions).

Specifications

Antigen Influenza B (virions)
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Influenza B, strain Yamagata.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Purified virions. Does not cross-react with influenza A virus, Parainfluenza virus (types 1-3), Respiratory Syncytial virus or Adenovirus. Negative against monkey kidney cells by indirect immunofluorescence.
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.