Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INO80, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89119493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-493 Supplier Abnova Corporation Supplier No. PAB17830
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of INO80.

INOC1 is the human homolog of S. cerevisiae Ino80, the catalytic ATPase subunit of the INO80 chromatin remodeling complex (Shen et al., 2000 [PubMed 10952318]; Bakshi et al., 2004 [PubMed 15207721]). INOC1 defines a subfamily of SWI2/SNF2 chromatin remodeling proteins; see SMARCA (MIM 600014) for background information.[supplied by OMIM

Sequence: C-TQEDYDSNHFKAQ

Specifications

Antigen INO80
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of INO80.
Dilution ELISA (1:4000) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene INO80
Gene Alias INO80A/INOC1/hINO80
Gene Symbols INO80
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human INO80.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54617
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.