Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF6, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132189 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-189 Supplier Abnova Corporation Supplier No. PAB6150
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of IRF6.

This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation. [provided by RefSeq

Sequence: C-TPSMQLPPALPPQ

Specifications

Antigen IRF6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of IRF6.
Dilution ELISA (1:32000) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene IRF6
Gene Alias LPS/OFC6/PIT/PPS/VWS
Gene Symbols IRF6
Host Species Goat
Immunogen A synthetic peptide corresponding to human IRF6.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3664
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.