Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITM2B, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-704 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-704 Supplier Abnova Corporation Supplier No. PAB15636
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of ITM2B.

Sequence: C-HDKETYKLQRRETIK

Specifications

Antigen ITM2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of ITM2B.
Dilution ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ITM2B
Gene Alias ABRI/BRI/BRI2/BRICD2B/E25B/E3-16/FBD
Gene Symbols ITM2B
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human ITM2B.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9445
Test Specificity Approximately 33 KDa band observed in human brain (cerebellum and hippocampus) lysates (calculated MW of 30.3 KDa according to NP_068839.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.