Learn More
RGS13, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RGS13.
Supplier: Abnova Corporation PAB19639
Description
The protein encoded by this gene is a member of the regulator of G protein signaling (RGS) family. RGS family members share similarity with S. cerevisiae SST2 and C. elegans egl-10 proteins, which contain a characteristic conserved RGS domain. RGS proteins accelerate GTPase activity of G protein alpha-subunits, thereby driving G protein into their inactive GDP-bound form, thus negatively regulating G protein signaling. RGS proteins have been implicated in the fine tuning of a variety of cellular events in response to G protein-coupled receptor activation. The biological function of this gene, however, is unknown. Two transcript variants encoding the same isoform exist. [provided by RefSeq
Sequence: DESKRPPSNLTLEE-CSpecifications
RGS13 | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of RGS13. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
MGC17173 | |
Goat | |
Antigen affinity purification | |
RUO | |
6003 | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.5-1ug/mL) The optimal working dilution should be determined by the end user. | |
RGS13 | |
RGS13 | |
A synthetic peptide corresponding to N-terminus of human RGS13. | |
100 μg | |
Primary | |
Reported variants represent identical proteins: NP_002918.1, NP_658912.1 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only