Learn More
LAMA3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of LAMA3.
Supplier: Abnova Corporation PAB6938
Description
Laminins are basement membrane components thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. The protein encoded by this gene is the alpha-3 subunit of laminin 5, which is a complex glycoprotein composed of three subunits (alpha, beta, and gamma). Laminin 5 is thought to be involved in cell adhesion, signal transduction and differentiation of keratinocytes. Mutations in this gene have been identified as the cause of Herlitz type junctional epidermolysis bullosa. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq
Sequence: NNQCHSSHKRRTKSpecifications
LAMA3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) The optimal working dilution should be determined by the end user. | |
LAMA3 | |
LAMA3 | |
A synthetic peptide corresponding to human LAMA3. | |
100 μg | |
Primary | |
This antibody is expected to recognize isoform 1 (NP_937762.1) only. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of LAMA3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
BM600/E170/LAMNA/LOCS/lama3a | |
Goat | |
Antigen affinity purification | |
RUO | |
3909 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only