Learn More
LARGE Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of LARGE.
Supplier: Abnova Corporation PAB18595
Description
This gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein. [provided by RefSeq
Sequence: C-SEADVNSENLQKQSpecifications
LARGE | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
LARGE | |
LARGE | |
A synthetic peptide corresponding to amino acids 421-433 at internal region of human LARGE. | |
100 μg | |
Primary | |
Reported variants represent identical protein: NP_004728.1, NP_598397.1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of LARGE. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
KIAA0609/MDC1D | |
Goat | |
Antigen affinity purification | |
RUO | |
9215 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only