Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEPR, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-896 Supplier Abnova Corporation Supplier No. PAB7037
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of LEPR.

Leptin (LEP; MIM 164160), an adipocyte-specific hormone that regulates adipose-tissue mass through hypothalamic effects on satiety and energy expenditure, acts through the leptin receptor (LEPR), a single-transmembrane-domain receptor of the cytokine receptor family.[supplied by OMIM

Sequence: C-TQDDIEKHQSDAG

Specifications

Antigen LEPR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of LEPR.
Dilution ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene LEPR
Gene Alias CD295/OBR
Gene Symbols LEPR
Host Species Goat
Immunogen A synthetic peptide corresponding to human LEPR.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3953
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_002294.2, NP_0010.03679.1, NP_0010.03680.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.