Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC494499 (GST Ya), Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116759 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-759 Supplier Abnova Corporation Supplier No. PAB13599
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native GST Ya.

Specifications

Antigen LOC494499 (GST Ya)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native GST Ya.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In purified antiserum
Gene LOC494499
Gene Alias MGC114217
Gene Symbols LOC494499
Host Species Goat
Immunogen Native purified GST Ya from rat liver.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494499
Test Specificity Goat antisera specific to Rat Glutathione S Transferase Ya. This antibody does not cross react with the mu or pi class of GST.
Target Species Human, Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.