Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC644096, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89118909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-909 Supplier Abnova Corporation Supplier No. PAB17244
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of LOC644096.

The succinate dehydrogenase (SDH) complex, or complex II of the mitochondrial respiratory chain, is composed of 4 individual subunits (see SDHA, MIM 600857). SDHAF1 is essential for SDH assembly but does not physically associate with the complex in vivo (Ghezzi et al., 2009).[supplied by OMIM

Sequence: C-HDSTGAPETRPDGR

Specifications

Antigen LOC644096
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of LOC644096.
Dilution ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene LOC644096
Gene Symbols LOC644096
Host Species Goat
Immunogen A synthetic peptide corresponding to human LOC644096.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644096
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.