Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly6e, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89120210 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-210 Supplier Abnova Corporation Supplier No. PAB18651
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of Ly6e.

Sequence: SCQEKDHYCIT

Specifications

Antigen Ly6e
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of Ly6e.
Dilution ELISA (1:16000) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene Ly6e
Gene Alias 9804/Ly67/RIG-E/Sca-2/TSA-1/Tsa1
Gene Symbols Ly6e
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of mouse Ly6e.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17069
Test Specificity Reported variants represent identical protein: NP_002337.1, NP_001120685.1.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.