Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MC5R, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89117713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-713 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-713 Supplier Abnova Corporation Supplier No. PAB15646
Add to Cart
Add to Cart

Goat polyclonal antibody raised aganist synthetic peptide of MC5R.

Sequence: PNVKNKSSPCED

Specifications

Antigen MC5R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised aganist synthetic peptide of MC5R.
Dilution ELISA (1:8000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene MC5R
Gene Symbols MC5R
Host Species Goat
Immunogen A synthetic peptide corresponding to human MC5R.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4161
Test Specificity Approximately 37 KDa band observed in human and mouse skeletal muscle lysates (calculated MW of 36.6 KDa according to NP_005904.1). An additional band of unknown identity was also consistently observed at 30KDa. This band was successfully blocked by incubation with the immunizing peptide.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.