Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mcpt9/Cma2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Mcpt9/Cma2.
Supplier: Abnova Corporation PAB11550
Description
Sequence: CKIFKHYKDSLQSpecifications
Mcpt9/Cma2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) The optimal working dilution should be determined by the end user. | |
MMCP-1/MMCP9 | |
Goat | |
Antigen affinity purification | |
RUO | |
17232/545055 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of Mcpt9/Cma2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
Mcpt9 | |
A synthetic peptide corresponding to mouse Mcpt9/Cma2. | |
100 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction