Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MON1A Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120185 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-185 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-185 Supplier Abnova Corporation Supplier No. PAB18629
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of MON1A.

Sequence: HISYLEPDTDLC

Specifications

Antigen MON1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of MON1A.
Dilution ELISA (1:128000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene MON1A
Gene Alias FLJ97088/MGC13272/SAND1
Gene Symbols MON1A
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 478-489 at internal region of human MON1A.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84315
Test Specificity This antibody is expected to recognize isoforms a and b (NP_115731.2; NP_001135973.1). Amino acid numbering in name refers to sequence NP_115731.2.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.