Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Heavy Chain Goat anti-Monkey, Texas Red, Polyclonal, Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP173561 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173561 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173561 Supplier Novus Biologicals Supplier No. NBP173561

Goat Polyclonal Antibody

IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.

Specifications

Antigen IgG Heavy Chain
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Texas Red
Dilution Western Blot, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Goat
Immunogen Goat anti-Monkey IgG (gamma chain) Secondary Antibody was produced by repeated immunization with monkey IgG gamma heavy chain in goat.
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Specificity was confirmed by ELISA at less than 1% cross reactivity against other Monkey heavy or light chain isotypes.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.