Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgE Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
SDP

Catalog No. NB7531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7531 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7531 Supplier Novus Biologicals Supplier No. NB7531

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Goat Polyclonal Antibody

IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen IgE Heavy Chain
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:1000 - 1:10000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Gene Symbols IGHE
Host Species Goat
Immunogen Mouse IgE
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3497
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with mouse IgE. This may cross react with IgE from other species.
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.