Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG2b Secondary Antibody, FITC (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NB002846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002846 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002846 Supplier Novus Biologicals Supplier No. NBP268514500UG

Goat Polyclonal Antibody

IgG2b Polyclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.

Specifications

Antigen IgG2b
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:1000, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence Neat - 1:10
Formulation PBS
Host Species Goat
Immunogen Mouse IgG2b paraproteins
Purification Method Affinity Purified
Quantity 500 μg
Test Specificity Goat anti Mouse IgG2b antibody recognizes Mouse IgG2b as has been pre-absorbed against mouse IgM, IgG1, IgG2a, IgG3 and IgA, pooled human sera and purified human paraproteins. The antibody shows minimal cross-reactivity with human immunoglobulins.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.