Learn More
Description
This gene encodes a mitochondrial inner membrane protein that is implicated in the metabolism of reactive oxygen species. Mutations in this gene have been associated with the hepatocerebral form of mitochondrial DNA depletion syndrome (MDDS). [provided by RefSeq
Specifications
Specifications
| Antigen | MPV17 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Description | Goat polyclonal antibody raised against synthetic peptide of MPV17. |
| Dilution | ELISA (1:32000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) |
| Gene | MPV17 |
| Gene Alias | SYM1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.