Learn More
MPZ, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MPZ.
Supplier: Abnova Corporation PAB7332
Description
This gene encodes a major structural protein of peripheral myelin. Mutations in this gene result in the autosomal dominant form of Charcot-Marie-Tooth disease type 1 as other polyneuropathies. [provided by RefSeq
Sequence: C-DHSRSTKAVSEKSpecifications
MPZ | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
MPZ | |
MPZ | |
A synthetic peptide corresponding to human MPZ. | |
100 μg | |
Primary | |
The presence of this PNS protein in Rat Spinal Cord is likely due to contamination with spinal roots during extraction from the animal. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of MPZ. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
CHM/CMT1/CMT1B/CMT2I/CMT2J/CMT4E/CMTDI3/DSS/HMSNIB/MPP/P0 | |
Goat | |
Antigen affinity purification | |
RUO | |
4359 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only