Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acylmannosamine-1-Dehydrogenase (NAM-DH) (Pseudomonas), Goat, Polyclonal Antibody, Abnova™

Catalog No. 89138432 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-432 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-432 Supplier Abnova Corporation Supplier No. PAB28047
Add to Cart
Add to Cart

Goat polyclonal antibody raised against full-length recombinant N-Acylmannosamine-1-Dehydrogenase (NAM-DH) (Pseudomonas).

Specifications

Antigen N-Acylmannosamine-1-Dehydrogenase (NAM-DH) (Pseudomonas)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against full length recombinant N-Acylmannosamine-1-Dehydrogenase (NAM-DH) (Pseudomonas).
Dilution ELISA (1:5000 - 1:20000) Immunoprecipitation (1:100) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Goat
Immunogen Recombinant protein corresponding to full length Pseudomonas NAM-DH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7614553
Target Species Bacteria
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.