Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCAM1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NCAM1.
Supplier: Abnova Corporation PAB19678
Description
Sequence: C-KASWTRPEKQETLDSpecifications
NCAM1 | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of NCAM1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
CD56/MSK39/NCAM | |
Goat | |
Antigen affinity purification | |
RUO | |
4684 | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
NCAM1 | |
NCAM1 | |
A synthetic peptide corresponding to internal region of human NCAM1. | |
100 μg | |
Primary | |
This antibody is expected to recognize reported isoform 2 (NP_851996.2) only. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction