Learn More
NDUFS1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NDUFS1.
Supplier: Abnova Corporation PAB18573
Description
The protein encoded by this gene belongs to the complex I 75kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur (IP) fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. [provided by RefSeq
Sequence: C-TEKSATYVNTEGRSpecifications
NDUFS1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
NDUFS1 | |
NDUFS1 | |
A synthetic peptide corresponding to amino acids at internal region of human NDUFS1. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of NDUFS1. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
CI-75Kd/MGC26839/PRO1304 | |
Goat | |
Antigen affinity purification | |
RUO | |
4719 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only