Learn More
NFIL3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NFIL3.
Supplier: Abnova Corporation PAB6068
Description
Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).[supplied by OMIM
Sequence: C-RLIATQPISASDSGSpecifications
NFIL3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
NFIL3 | |
NFIL3 | |
A synthetic peptide corresponding to human NFIL3. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of NFIL3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
E4BP4/IL3BP1/NF-IL3A/NFIL3A | |
Goat | |
Antigen affinity purification | |
RUO | |
4783 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only