Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1H3/NR1H2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-604 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-604 Supplier Abnova Corporation Supplier No. PAB6691
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.

Sequence: CRLQDKKLPPLLSEI

Specifications

Antigen NR1H3/NR1H2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.
Dilution ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Alias LXR-a/LXRA/RLD-1
Gene Symbols NR1H3
Host Species Goat
Immunogen A synthetic peptide corresponding to human NR1H3, NR1H2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062/7376
Test Specificity This antibody is expected to recognize an epitope corresponding to aa 429-442 of human LXR alpha protein (NP_005684) and to aa 443-456 of human LXR beta protein (NP_009052).
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.