Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRXN1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-388 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-388 Supplier Abnova Corporation Supplier No. PAB7583
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of NRXN1.

Neurexins function in the vertebrate nervous system as cell adhesion molecules and receptors. Two neurexin genes are among the largest known in human (NRXN1 and NRXN3). By using alternate promoters, splice sites and exons, predictions of hundreds or even thousands of distinct mRNAs have been made. Most transcripts use the upstream promoter and encode alpha-neurexin isoforms; fewer transcripts are produced from the downstream promoter and encode beta-neurexin isoforms. Alpha-neurexins contain epidermal growth factor-like (EGF-like) sequences and laminin G domains, and they interact with neurexophilins. Beta-neurexins lack EGF-like sequences and contain fewer laminin G domains than alpha-neurexins. The RefSeq Project has decided to create only a few representative transcript variants of the multitude that are possible. [provided by RefSeq

Sequence: C-KEKQPSSAKSSNKN

Specifications

Antigen NRXN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of NRXN1.
Dilution ELISA (1:32000) Western Blot (0.05-0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene NRXN1
Gene Alias DKFZp313P2036/FLJ35941/Hs.22998/KIAA0578
Gene Symbols NRXN1
Host Species Goat
Immunogen A synthetic peptide corresponding to human NRXN1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9378
Test Specificity This antibody is expected to recognize both reported isoforms (NP_004792.1 and NP_620072.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.