Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP85, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132483 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-483 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-483 Supplier Abnova Corporation Supplier No. PAB6540
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of NUP85.

Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160. [provided by RefSeq

Sequence: C-ARAIIREGSLEGS

Specifications

Antigen NUP85
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of NUP85.
Dilution ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene NUP85
Gene Alias FLJ12549/Nup75
Gene Symbols NUP85
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human NUP85.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79902
Test Specificity In contrast to other anti-Pericentrin antibodies available, this antibody is NOT expected to recognize Kendrin (also known as Pericentrin 2). This may explain why it recognizes a 75 KDa band (the expected size according to NP_079120) and not a 220 KDa band (as reported for other anti-pericentrin antibodies).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.