Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL3, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132465 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-465 Supplier Abnova Corporation Supplier No. PAB6515
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of OSBPL3.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Several transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MSDEKNLGVSQKL-C

Specifications

Antigen OSBPL3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of OSBPL3.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene OSBPL3
Gene Alias DKFZp667P1518/KIAA0704/MGC21526/ORP-3/ORP3/OSBP3
Gene Symbols OSBPL3
Host Species Goat
Immunogen A synthetic peptide corresponding to human OSBPL3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26031
Test Specificity This antibody is expected to recognize all six human isoforms of this protein.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.