Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL9, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132469 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-469 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-469 Supplier Abnova Corporation Supplier No. PAB6520
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of OSBPL9.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Multiple transcript variants, most of which encode distinct isoforms, have been identified. [provided by RefSeq

Sequence: C-EPLLKRLGAAKH

Specifications

Antigen OSBPL9
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of OSBPL9.
Dilution ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene OSBPL9
Gene Alias FLJ12492/FLJ14629/FLJ14801/FLJ32055/FLJ34384/MGC15035/ORP9
Gene Symbols OSBPL9
Host Species Goat
Immunogen A synthetic peptide corresponding to human OSBPL9.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114883
Test Specificity This antibody is expected to recognize all six human isoforms of this protein (NP_683702.1 and NP_68370.3.1 are varients which represent the same isoform).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.