Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAPD5 Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89121182 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-182 Supplier Abnova Corporation Supplier No. PAB19656
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of PAPD5.

Sequence: C-HALPAEQRDFLP

Specifications

Antigen PAPD5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of PAPD5.
Dilution Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene PAPD5
Gene Alias FLJ40270/TRF4-2
Gene Symbols PAPD5
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 78-90 at internal region of human PAPD5.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64282
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001035374.2; NP_001035375.2).
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.