Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parainfluenza 1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137803 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-803 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-803 Supplier Abnova Corporation Supplier No. PAB14020
Add to Cart
Add to Cart

Goat polyclonal antibody raised against Parainfluenza 1.

Specifications

Antigen Parainfluenza 1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against Parainfluenza 1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Parainfluenza Virus Type 1 Cantell strain.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Parainfluenza 1, all structural antigens. Recognizes Sendai virus by ELISA. Does not cross-react with Parainfluenza types 2 and 3, Influenza A, Influenza B, Respiratory Syncytial Virus, HSV1, HSV2, Adenovirus, CMV, Measles, Mumps and Rubella by indirect IFA. Does not react with HEp-2 cells or monkey kidney cells by indirect IFA.
Target Species Influenza virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.