Learn More
PARD6A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PARD6A.
Supplier: Abnova Corporation PAB6553
Description
This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition (EMT) that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: C-GSRIRGDGSGFSLSpecifications
PARD6A | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Immunohistochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
PARD6A | |
PARD6A | |
A synthetic peptide corresponding to human PARD6A. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PARD6A. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
PAR-6A/PAR6/PAR6C/PAR6alpha/TAX40/TIP-40 | |
Goat | |
Antigen affinity purification | |
RUO | |
50855 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only