Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARK2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116886 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-886 Supplier Abnova Corporation Supplier No. PAB14022

Goat polyclonal antibody raised against synthetic peptide of PARK2.

The precise function of this gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support. [provided by RefSeq

Sequence: TGGDDPRNAAGGCER

Specifications

Antigen PARK2
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of PARK2.
Dilution ELISA (>1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (>1:300) Western Blot (>1:5000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene PARK2
Gene Alias AR-JP/LPRS2/PDJ/PRKN
Gene Symbols PARK2
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 83-97 of human PARK2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5071
Test Specificity Amino acid sequence 83-97 of human parkin protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.