Learn More
PAX8 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PAX8.
Supplier: Abnova Corporation PAB6201
Description
This gene encodes a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternateively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq
Sequence: PHNSIRSGHGGLN-CSpecifications
PAX8 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Immunohistochemistry (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
PAX8 | |
Goat | |
Antigen affinity purification | |
RUO | |
7849 | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PAX8. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
PAX8 | |
A synthetic peptide corresponding to human PAX8. | |
100 μg | |
Primary | |
This antibody is expected to recognize all five human isoforms (as represented by NP_0.03457.1, NP_0.39245.1, NP_0.39246.1, NP_0.39247.1, NP_054698.1) | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only