Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNX, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PCNX.
Supplier: Abnova Corporation PAB6003
Description
Sequence: C-TVIETGVLELGAEVSpecifications
PCNX | |
Polyclonal | |
Unconjugated | |
ELISA (1:64000) The optimal working dilution should be determined by the end user. | |
PCNX | |
PCNX | |
A synthetic peptide corresponding to human PCNX. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PCNX. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
FLJ23409/FLJ45663/KIAA0805/KIAA0995/PCNXL1 | |
Goat | |
Antigen affinity purification | |
RUO | |
22990 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction